0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
α,α,α,α',α',α'-Hexafluoro-o-xylene
α,α,α,α',α',α'-Hexafluoro-o-xylene 97%
α,α,α,α',α',α'-Hexafluoro-o-xylene 97%
α,α,α,α',α',α'-Hexafluoro-o-xylene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
α,α,α,α',α',α'-Hexafluoro-o-xylene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
433-95-4
:
Formula:
C₈H₄F₆
MDL Number:
MFCD03094218
Storage Temperature:
Ambient