0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3,5-Bis(trifluoromethyl)benzamidoxime
3,5-Bis(trifluoromethyl)benzamidoxime 97%
3,5-Bis(trifluoromethyl)benzamidoxime 97%
:
Apollo Scientific
3,5-Bis(trifluoromethyl)benzamidoxime 97%
:
Apollo Scientific
:
72111-09-2
:
:
C₉H₆F₆N₂O
:
MFCD00068188
:
272.15 g/mol
:
131…133 °C
:
GHS07
:
Ambient