0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-Fluoro-2,6-dinitrophenol
4-Fluoro-2,6-dinitrophenol 98%
4-Fluoro-2,6-dinitrophenol 98%
Irritant
4-Fluoro-2,6-dinitrophenol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Fluoro-2,6-dinitrophenol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
364-32-9
:
Formula:
C₆H₃FN₂O₅
MDL Number:
MFCD00179333
:
202.1 g/mol
Melting Pt:
54…56 °C
:
GHS07
Storage Temperature:
Ambient