


3,4-Dinitrofluorobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:364-53-4
:
Formula:C₆H₃FN₂O₄
MDL Number:MFCD00236671
Melting Pt:55…56 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,II
:186.1 g/mol
:GHS05
:GHS06
:GHS08