Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Ethoxy-2,6-difluoroaniline
4-Ethoxy-2,6-difluoroaniline 97%
4-Ethoxy-2,6-difluoroaniline 97%
4-Ethoxy-2,6-difluoroaniline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Ethoxy-2,6-difluoroaniline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1017779-79-1
:
Formula:
C₈H₉F₂NO
MDL Number:
MFCD09832394
:
173.16 g/mol
Storage Temperature:
Ambient