Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-2,3-difluoroaniline
6-Chloro-2,3-difluoroaniline 98%
6-Chloro-2,3-difluoroaniline 98%
Irritant
6-Chloro-2,3-difluoroaniline 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Chloro-2,3-difluoroaniline 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1373920-77-4
:
Formula:
C₆H₄ClF₂N
MDL Number:
MFCD22201032
:
163.55 g/mol
:
GHS07
Storage Temperature:
Ambient