Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-2,3-difluorobenzaldehyde
6-Bromo-2,3-difluorobenzaldehyde 99%
6-Bromo-2,3-difluorobenzaldehyde 99%
Irritant
6-Bromo-2,3-difluorobenzaldehyde 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Bromo-2,3-difluorobenzaldehyde 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
360576-04-1
:
Formula:
C₇H₃BrF₂O
MDL Number:
MFCD07368119
:
221 g/mol
Melting Pt:
39…43 °C
:
GHS07
Storage Temperature:
Ambient