
4,5-Difluoro-2-methoxybenzyl bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886499-64-5
:
Formula:C₈H₇BrF₂O
MDL Number:MFCD06660220
:237.04 g/mol
Melting Pt:40…43 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05