Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2-methoxyaniline
4,5-Difluoro-2-methoxyaniline 97%
4,5-Difluoro-2-methoxyaniline 97%
Irritant
4,5-Difluoro-2-methoxyaniline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Difluoro-2-methoxyaniline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1017779-71-3
:
Formula:
C₇H₇F₂NO
MDL Number:
MFCD09832387
:
159.14 g/mol
Storage Temperature:
Ambient
UN:
2810
ADR:
6.1,III
:
GHS07