
2,3-Difluoro-6-methoxybenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:221202-34-2
:
Formula:C₈H₅F₂NO
MDL Number:MFCD04116019
:169.13 g/mol
Melting Pt:79…82 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06