0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,6-Dichloro-3-(trifluoromethyl)benzamide
2,6-Dichloro-3-(trifluoromethyl)benzamide 98%
2,6-Dichloro-3-(trifluoromethyl)benzamide 98%
2,6-Dichloro-3-(trifluoromethyl)benzamide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,6-Dichloro-3-(trifluoromethyl)benzamide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
53012-81-0
:
Formula:
C₈H₄Cl₂F₃NO
MDL Number:
MFCD11519346
Melting Pt:
125…128 °C
Storage Temperature:
Ambient