Order Entry
Export
ContactUsLinkComponent
3-Ethoxy-2,4-difluorobenzylamine 95%
3-Ethoxy-2,4-difluorobenzylamine 95%
Catalog # APOSPC302775-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Ethoxy-2,4-difluorobenzylamine 95%
Catalog # APOSPC302775-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:1017778-36-7
Formula:C₉H₁₁F₂NO
MDL Number:MFCD09258733