Order Entry
Export
ContactUsLinkComponent
6-Chloro-2,3-difluorobenzoyl chloride 98%
6-Chloro-2,3-difluorobenzoyl chloride 98%
Catalog # APOSPC302763-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-2,3-difluorobenzoyl chloride 98%
Catalog # APOSPC302763-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211 g/mol
Storage Temperature:Ambient
CAS Number:1208078-25-4
Formula:C₇H₂Cl₂F₂O
MDL Number:MFCD12922578