
3,6-Dichloro-2-fluorobenzyl bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916420-69-4
:
Formula:C₇H₄BrCl₂F
MDL Number:MFCD09025389
Melting Pt:42…45 °C
UN:3261
ADR:8,II
:257.92 g/mol
:GHS05
Storage Temperature:Ambient