Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde 97%
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde 97%
Irritant
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
180134-14-9
:
Formula:
C₁₀H₆F₆O₂
MDL Number:
MFCD06660316
:
272.15 g/mol
Melting Pt:
36…39 °C
:
GHS07
Storage Temperature:
Ambient