Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethoxy-3,5-difluorobenzaldehyde
2-Ethoxy-3,5-difluorobenzaldehyde 98%
2-Ethoxy-3,5-difluorobenzaldehyde 98%
2-Ethoxy-3,5-difluorobenzaldehyde 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Ethoxy-3,5-difluorobenzaldehyde 98%
Supplier:
Apollo Scientific
About this item
:
Formula:
C₉H₈F₂O₂
MDL Number:
MFCD06247321
:
186.16 g/mol
Storage Temperature:
Ambient