Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichloro-6-fluoroaniline
2,3-Dichloro-6-fluoroaniline 97%
2,3-Dichloro-6-fluoroaniline 97%
2,3-Dichloro-6-fluoroaniline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Dichloro-6-fluoroaniline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1017777-51-3
:
Formula:
C₇H₄Cl₂F₃N
MDL Number:
MFCD09832261
:
180.01 g/mol
Melting Pt:
52…54 °C
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III