Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethoxy-3,5-difluorobenzylamine
2-Ethoxy-3,5-difluorobenzylamine 97%
2-Ethoxy-3,5-difluorobenzylamine 97%
2-Ethoxy-3,5-difluorobenzylamine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Ethoxy-3,5-difluorobenzylamine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1017779-86-0
:
Formula:
C₉H₁₁F₂NO
MDL Number:
MFCD09258721
:
187.19 g/mol
Storage Temperature:
Ambient