Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Chloro-4,5-difluoroaniline

Irritant
Health
Supplier:   Apollo Scientific
2-Chloro-4,5-difluoroaniline
Supplier:  Apollo Scientific
Certificates
2-Chloro-4,5-difluoroaniline

3 Options Available Below

Product Details & Documents
  • CAS Number:2613-32-3
  • Formula:C₆H₄ClF₂N
  • MDL Number:MFCD08448325
  • Molecular Weight:163.55 g/mol
  • Melting Pt:55…58 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 52,10 €
    52,10 €
    1 * 25 g
       52,10 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 168,00 €
    168,00 €
    1 * 100 g
       168,00 €