Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoro-3-methoxybenzaldehyde
2,4-Difluoro-3-methoxybenzaldehyde 90%
2,4-Difluoro-3-methoxybenzaldehyde 90%
Irritant
2,4-Difluoro-3-methoxybenzaldehyde 90%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Difluoro-3-methoxybenzaldehyde 90%
Supplier:
Apollo Scientific
About this item
CAS Number:
870837-66-4
:
Formula:
C₈H₆F₂O₂
MDL Number:
MFCD04115930
:
172.13 g/mol
:
GHS07
Storage Temperature:
Ambient