Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichloro-6-fluorophenol
2,3-Dichloro-6-fluorophenol 98%
2,3-Dichloro-6-fluorophenol 98%
Irritant
2,3-Dichloro-6-fluorophenol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Dichloro-6-fluorophenol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
886497-60-5
:
Formula:
C₆H₃Cl₂FO
:
180.99 g/mol
:
GHS07
Storage Temperature:
Ambient