Order Entry
Export
ContactUsLinkComponent
6-Chloro-2,3-difluorophenylacetonitrile 97%
6-Chloro-2,3-difluorophenylacetonitrile 97%
Catalog # APOSPC302366-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-2,3-difluorophenylacetonitrile 97%
Catalog # APOSPC302366-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.57 g/mol
Storage Temperature:Ambient
CAS Number:1373920-84-3
Formula:C₈H₄ClF₂N
MDL Number:MFCD22201041