3,4-Difluoro-5-methylbenzaldehyde
Catalog # APOSPC302269-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:156.13 g/mol
Storage Temperature:Ambient
CAS Number:1017778-64-1
Formula:C₈H₆F₂O
MDL Number:MFCD09832333