Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethoxy-3,5-difluorophenol
2-Ethoxy-3,5-difluorophenol 98%
2-Ethoxy-3,5-difluorophenol 98%
2-Ethoxy-3,5-difluorophenol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Ethoxy-3,5-difluorophenol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1017779-78-0
:
Formula:
C₈H₈F₂O₂
MDL Number:
MFCD09258715
Storage Temperature:
Ambient