

3,5-Dimethyl-4-fluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1840-27-3
:
Formula:C₈H₁₀FN
MDL Number:MFCD09832247
Melting Pt:30…32 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:139.17 g/mol
:GHS07
:GHS08