4,6-Difluoroindole-2-carboxylic acid 97%
Catalog # APOSPC3022-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:197.1400 g/mol
Storage Temperature:Ambient
CAS Number:247564-66-5
Formula:C₉H₅F₂NO₂
MDL Number:MFCD01863158