0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3,6-Dichloro-2-fluorobenzoic acid
3,6-Dichloro-2-fluorobenzoic acid 98%
3,6-Dichloro-2-fluorobenzoic acid 98%
:
Apollo Scientific
3,6-Dichloro-2-fluorobenzoic acid 98%
:
Apollo Scientific
:
916420-62-7
:
:
C₇H₃Cl₂FO₂
:
MFCD09025382
:
209 g/mol
:
150…153 °C
:
GHS07
:
Ambient