
2,3-Difluoro-4-methylbenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:508203-48-3
:
Formula:C₈H₅F₂N
MDL Number:MFCD06660243
:153.13 g/mol
Melting Pt:43…45 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07