Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-5-fluorobenzylamine
2,4-Dichloro-5-fluorobenzylamine 97%
2,4-Dichloro-5-fluorobenzylamine 97%
2,4-Dichloro-5-fluorobenzylamine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dichloro-5-fluorobenzylamine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
939980-28-6
:
Formula:
C₇H₆Cl₂FN
MDL Number:
MFCD06660211
Storage Temperature:
Ambient