1-(3,4-Difluorobenzyl)-1H-pyrrole
Catalog # APOSPC300001-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:193.2 g/mol
Storage Temperature:Ambient
CAS Number:1247669-63-1
Formula:C₁₁H₉F₂N
MDL Number:MFCD14684087