1,6-Diiodoperfluorohexane 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Perfluoro-1,6-diiodohexane
- CAS Number:375-80-4
- Formula:C₆F₁₂I₂
- MDL Number:MFCD00042264
- Molecular Weight:553.86 g/mol
- Boiling Pt:114…116 °C/100mmHg
- Melting Pt:25 °C
- Density:2.357 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- EINECS:206-794-7
3 Options Available Below
Product Details & Documents
Synonyms: Perfluoro-1,6-diiodohexane
- CAS Number:375-80-4
- Formula:C₆F₁₂I₂
- MDL Number:MFCD00042264
- Molecular Weight:553.86 g/mol
- Boiling Pt:114…116 °C/100mmHg
- Melting Pt:25 °C
- Density:2.357 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- EINECS:206-794-7



