
1,4-Diiodoperfluorobutane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:375-50-8
:
Formula:C₄F₈I₂
MDL Number:MFCD00042263
:453.84 g/mol
Boiling Pt:150 °C
Melting Pt:–9 °C
Density:2.474 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:206-788-4
:GHS07