

2,6-Difluoropyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1513-65-1
:
Formula:C₅H₃F₂N
MDL Number:MFCD00006227
:115.08 g/mol
Boiling Pt:125 °C
Density:1.268 g/cm³ (25 °C)
Flash Pt:33 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07