Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Difluoropropane 97%

1,3-Difluoropropane 97%

Irritant
Supplier:   Apollo Scientific
1,3-Difluoropropane 97%
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:462-39-5
  • Formula:C₃H₆F₂
  • MDL Number:MFCD00040969
  • Molecular Weight:80.08 g/mol
  • Boiling Pt:40…42 °C
  • Density:1.006 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 267,00 €
    267,00 €
    1 * 5 g
       267,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSPC2877B-5G
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 436,00 €
    436,00 €
    1 * 25 g
       436,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSPC2877B-25G