Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluorophenyl isocyanate
2,4-Difluorophenyl isocyanate 98%
2,4-Difluorophenyl isocyanate 98%
:
Apollo Scientific
2,4-Difluorophenyl isocyanate 98%
:
Apollo Scientific
:
59025-55-7
:
:
C₇H₃F₂NO
:
MFCD00001997
:
42 °C
:
1.309 g/cm³ (25 °C)
:
55 °C
:
2478
:
3+6.1,III
:
155.1 g/mol
:
GHS02
:
GHS07
:
GHS08
:
Refrigerator