Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluoro-DL-phenylalanine
3,4-Difluoro-DL-phenylalanine 97%
3,4-Difluoro-DL-phenylalanine 97%
:
Apollo Scientific
3,4-Difluoro-DL-phenylalanine 97%
:
Apollo Scientific
:
32133-36-1
:
:
C₉H₉F₂NO₂
:
MFCD00061307
:
237…240 °C
:
201.18 g/mol
:
GHS07
:
Ambient