
(2,5-Difluorophenyl)acetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69584-87-8
:
Formula:C₈H₅F₂N
MDL Number:MFCD00009972
:153.13 g/mol
Boiling Pt:90 °C/5mmHg
Density:1.233 g/cm³ (25 °C)
Flash Pt:103 °C
Storage Temperature:Ambient
EINECS:274-051-4
UN:3276
ADR:6.1,III
:GHS06