Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2,3-Difluorophenyl)acetonitrile
(2,3-Difluorophenyl)acetonitrile 97%
(2,3-Difluorophenyl)acetonitrile 97%
:
Apollo Scientific
(2,3-Difluorophenyl)acetonitrile 97%
:
Apollo Scientific
:
145689-34-5
:
:
C₈H₅F₂N
:
MFCD00061277
:
153.13 g/mol
:
1.2 g/cm³ (25 °C)
:
85 °C
:
Ambient
:
3276
:
6.1,III
:
GHS07