


3,5-Difluorophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2713-34-0
:
Formula:C₆H₄F₂O
MDL Number:MFCD00002255
Boiling Pt:65…68 °C/1mmHg
Melting Pt:54…57 °C
Flash Pt:71 °C
UN:2925
ADR:4.1+8,II
:130.09 g/mol
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient