

2,5-Difluorophenol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2713-31-7
:
Formula:C₆H₄F₂O
MDL Number:MFCD00042501
:130.09 g/mol
Boiling Pt:145 °C
Melting Pt:36…42 °C
Flash Pt:53 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,II
:GHS02
:GHS07