Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Difluorophenol 98%

2,3-Difluorophenol 98%

Flammable
Irritant
Supplier:   Apollo Scientific
2,3-Difluorophenol 98%
Supplier:  Apollo Scientific
Certificates
2,3-Difluorophenol 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:6418-38-8
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00010262
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:53…55 °C/25mmHg
  • Melting Pt:34…38 °C
  • Flash Pt:56 °C
  • Storage Temperature:Ambient
  • UN:1325
  • ADR:4.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 39,10 €
    39,10 €
    1 * 25 g
       39,10 €