

ω-Bromo-3,4-difluoroacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40706-98-7
:
Formula:C₈H₅BrF₂O
MDL Number:MFCD00085010
:235.03 g/mol
Melting Pt:34…35 °C
UN:3261
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Refrigerator