Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluoro-5-nitrobenzotrifluoride
3,4-Difluoro-5-nitrobenzotrifluoride 97%
3,4-Difluoro-5-nitrobenzotrifluoride 97%
3,4-Difluoro-5-nitrobenzotrifluoride 97%
Catalog # APOSPC2863E-1G
Supplier:
Apollo Scientific
CAS Number:
Expand image
3,4-Difluoro-5-nitrobenzotrifluoride 97%
Catalog # APOSPC2863E-1G
Supplier:
Apollo Scientific
CAS Number:
About this item
Product Details
Product Details