Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-2,4-Difluoromandelic acid
(±)-2,4-Difluoromandelic acid
(±)-2,4-Difluoromandelic acid
Irritant
(±)-2,4-Difluoromandelic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
(±)-2,4-Difluoromandelic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
132741-30-1
:
Formula:
C₈H₆F₂O₃
MDL Number:
MFCD00061296
:
188.13 g/mol
Melting Pt:
122…124 °C
:
GHS07
Storage Temperature:
Ambient