
1,2-Difluoro-4-iodobenzene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64248-58-4
:
Formula:C₆H₃F₂I
MDL Number:MFCD00044630
:239.99 g/mol
Boiling Pt:177…178 °C
Density:1.99 g/cm³ (25 °C)
Flash Pt:72 °C
:GHS07
Storage Temperature:Refrigerator