
1,3-Difluoro-4-iodobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2265-93-2
:
Formula:C₆H₃F₂I
MDL Number:MFCD00040151
:239.99 g/mol
Boiling Pt:175…176 °C
Density:2.006 g/cm³ (25 °C)
Flash Pt:67 °C
:GHS07
Storage Temperature:Refrigerator