

1,1-Difluoroethylene 99%
Catalog # APOSPC2810-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Boiling Pt:–83 °C
Boiling Pt:–83 °C
Melting Pt:–144 °C
Melting Pt:–144 °C
Density:0.617 g/cm³ (25 °C)
Flash Pt:<–60 °C
Flash Pt:<–60 °C
Storage Temperature:Ambient
UN:1959
ADR:2.1,
:64.03 g/mol
:GHS02
:GHS04
CAS Number:75-38-7
Formula:C₂H₂F₂
MDL Number:MFCD00000448