

2,2-Difluoroethanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:359-13-7
:
Formula:C₂H₄F₂O
MDL Number:MFCD00042248
:82.05 g/mol
Boiling Pt:95…96 °C
Melting Pt:–28 °C
Density:1.296 g/cm³ (25 °C)
Flash Pt:3 °C
Storage Temperature:Ambient
UN:1987
ADR:3,II
:GHS02
:GHS07