Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2-nitrophenol
4,5-Difluoro-2-nitrophenol 97%
4,5-Difluoro-2-nitrophenol 97%
Irritant
4,5-Difluoro-2-nitrophenol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Difluoro-2-nitrophenol 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
55346-97-9
:
Formula:
C₆H₃F₂NO₃
MDL Number:
MFCD03094187
:
175.0897 g/mol
Melting Pt:
54…57 °C
:
GHS07
Storage Temperature:
Ambient