Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluorocinnamic acid
2,3-Difluorocinnamic acid 97%
2,3-Difluorocinnamic acid 97%
Irritant
2,3-Difluorocinnamic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Difluorocinnamic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
207981-48-4
:
Formula:
C₉H₆F₂O₂
MDL Number:
MFCD00061291
:
184.14 g/mol
Melting Pt:
200…202 °C
:
GHS07
Storage Temperature:
Ambient